Learn More
Strem, An Ascensus Company CAS# 14282-91-8. 250mg. Hexaammineruthenium(III) chloride, 99%. MFCD00011478
SDP
Catalog No. 502016735
Encompass
CAS# 14282-91-8. 250mg. Hexaammineruthenium(III) chloride, 99%. MFCD00011478. Molecular Weight: 309.61. Molecular Formula: Ru(NH3)6Cl3. Color/form: pale yellow pwdr. Strem# 44-0620. http://www.strem.com/catalog/v/44-0620/
Catalog No. 50-201-6735 Supplier Strem, An Ascensus Company Supplier No. 440620250MG
CAS# 14282-91-8. 250mg. Hexaammineruthenium(III) chloride, 99%. MFCD00011478. Molecular Weight: 309.61. Molecular Formula: Ru(NH3)6Cl3. Color/form: pale yellow pwdr. Strem# 44-0620. http://www.strem.com/catalog/v/44-0620/