Learn More
Cayman Chemical RU-SKI 201 (hydrochloride), Purity: ≥95%, CAS: 2320262-09-5, 10 mg
SDP
Catalog No. 502983468
Encompass_Preferred
An Hhat inhibitor (IC50 = 0.2 μM for recombinant enzyme); inhibits palmitoylation of Shh in Shh-overexpressing HEK293 cells with a tagging IC50 (TC50) value of 0.87 μM; inhibits Hh signaling in H520, PANC-1, and MCF-7 cancer cells co-cultured with Shh-Light2 cells (IC50s = 4.8, 7.8, and 8.5 μM, respectively)
Catalog No. 50-298-3468 Supplier Cayman Chemical Supplier No. 2110110 MG
An Hhat inhibitor (IC50 = 0.2 μM for recombinant enzyme); inhibits palmitoylation of Shh in Shh-overexpressing HEK293 cells with a tagging IC50 (TC50) value of 0.87 μM; inhibits Hh signaling in H520, PANC-1, and MCF-7 cancer cells co-cultured with Shh-Light2 cells (IC50s = 4.8, 7.8, and 8.5 μM, respectively)