Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RU-SKI 43 hydrochloride | 1782573-67-4 | 98.81% | 423.01 | 1 ML
SDP

Catalog No. 5000387930
Encompass_Preferred

RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. It reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. This compound exhibits anti-cancer activity and is for research use only.

  • Potent and selective Hhat inhibitor
  • Reduces Gli-1 activation through Smoothened-independent non-canonical signaling
  • Decreases Akt and mTOR pathway activity
  • Exhibits anti-cancer activity
  • Purity of 98.81%
  • Available in solid form
  • Soluble in DMSO (≥ 51 mg/mL) and H2O (2.5 mg/mL)

Catalog No. 50-003-87930 Supplier Medchemexpress LLC Supplier No. HY18366A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RU-SKI 43 hydrochloride is a potent and selective Hedgehog acyltransferase (Hhat) inhibitor with an IC50 of 850 nM. It reduces Gli-1 activation through Smoothened-independent non-canonical signaling and decreases Akt and mTOR pathway activity. This compound exhibits anti-cancer activity and is for research use only.

  • Potent and selective Hhat inhibitor
  • Reduces Gli-1 activation through Smoothened-independent non-canonical signaling
  • Decreases Akt and mTOR pathway activity
  • Exhibits anti-cancer activity
  • Purity of 98.81%
  • Available in solid form
  • Soluble in DMSO (≥ 51 mg/mL) and H2O (2.5 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.