Learn More
Cayman Chemical RU-SKI 43hydrochlorIde 10mg
SDP
Catalog No. 502967029
Encompass_Preferred
A cell-permeable inhibitor of Hhat (IC50 = 0.85 µM); blocks palmitoylation of Shh without affecting palmitoylation of H-Ras or Fyn, myristoylation of c-Src, or acylation of Wnt3a; inhibits both autocrine and paracrine Shh-induced activation of Gli-mediated transcription
Catalog No. 50-296-7029 Supplier Cayman Chemical Supplier No. 1697210 MG
A cell-permeable inhibitor of Hhat (IC50 = 0.85 µM); blocks palmitoylation of Shh without affecting palmitoylation of H-Ras or Fyn, myristoylation of c-Src, or acylation of Wnt3a; inhibits both autocrine and paracrine Shh-induced activation of Gli-mediated transcription