Learn More
TARGETMOL CHEMICALS INC RU-TRAAK-2 5MG
SDP
Catalog No. 502579684
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RU-TRAAK-2 is a reversible inhibitor of TWIK-related arachidonic acid-stimulated K+ channel with no activity for non-K2P channels including Kv1.2 GIRK2 and Slo1. purity: 99%
Catalog No. 50-257-9684 Supplier TARGETMOL CHEMICALS INC Supplier No. T600615MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RU-TRAAK-2 is a reversible inhibitor of TWIK-related arachidonic acid-stimulated K+ channel with no activity for non-K2P channels including Kv1.2 GIRK2 and Slo1. purity: 99%