Learn More
Electron Microscopy Sciences EMS High Precision Rubber Grip Cutting Tweezer Style 15A, Angled Blade, 120 mm OAL
SDP
Catalog No. 501931005
Encompass_Preferred
Soft ESD safe ergonomic cushion grips (resistivity 10^8-10^9 Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Cutting tweezers for soft copper, gold, silver, or magnetic wires.
Blade: Angled
OAL: 120 mm
Catalog No. 50-193-1005 Supplier Electron Microscopy Sciences Supplier No. 7832509
Soft ESD safe ergonomic cushion grips (resistivity 10^8-10^9 Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Cutting tweezers for soft copper, gold, silver, or magnetic wires.
Blade: Angled
OAL: 120 mm