Learn More
Fritsch Milling And Sizing Inc Rubber Ball 20 mm Sieving Aid
SDP
Catalog No. 501534162
Encompass
1 rubber ball 20 mm dia. 
Is recommended as sieving aids for dry sieving of materials with a high proportion of fine particles.
Recommended quantity per sieve: 5 pieces
Catalog No. 50-153-4162 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.0180.15
Add to Cart
Add to Cart
1 rubber ball 20 mm dia. 
Is recommended as sieving aids for dry sieving of materials with a high proportion of fine particles.
Recommended quantity per sieve: 5 pieces