Learn More
Ted Pella Inc RUBBER BLOWER WITH PLASTIC TIP
SDP
Catalog No. NC2072773
Encompass
This Rubber Blower has a metal nozzle and inlet valve. The nozzle is plastic-sheathed to prevent marking of delicate parts. Provides a strong, directed air stream for cleaning when the bulb is squeezed. A substitute for pressurized cans. Swiss made.
Catalog No. NC2072773 Supplier Ted Pella Inc Supplier No. 54533
This Rubber Blower has a metal nozzle and inlet valve. The nozzle is plastic-sheathed to prevent marking of delicate parts. Provides a strong, directed air stream for cleaning when the bulb is squeezed. A substitute for pressurized cans. Swiss made.