Learn More
Andwin Scientific RUBBER CEMENT
SDP
Catalog No. 50949105
Encompass_Preferred
The rubber cement is an ideal adhesive for paper pastings and mountings of illustrations, drawings, and photographs. This product can also stick speciments onto SEM Stubs. If needed, the cement can be taken off with no difficulty. This product is sold individually.
Catalog No. 50-949-105 Supplier Andwin Scientific Supplier No. 72170
The rubber cement is an ideal adhesive for paper pastings and mountings of illustrations, drawings, and photographs. This product can also stick speciments onto SEM Stubs. If needed, the cement can be taken off with no difficulty. This product is sold individually.