Learn More
OHAUS Rubber Cover for 3 in. Platform
Catalog No. 01804428 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-428 Vortex Attachment
1 options
Catalog No. 01-804-428 Supplier OHAUS Supplier No. 83041442
Add to Cart
Add to Cart
Replacement rubber cover for the 3 in. (76 mm) platform (Part No 0K-0500-901).
  • This may be used on the Vortex Genie 1 Vortex Genie Pulse and the Vortex Genie 2 family of mixers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers