Learn More
Electron Microscopy Sciences EMS High Precision Rubber Grip Cutting Tweezer Style 15AGW, Predominantly angled Blade, 115 mm OAL
SDP
Catalog No. 501931006
Encompass_Preferred
Soft ESD safe ergonomic cushion grips (resistivity 10^8-10^9 Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Cutting tweezers for soft copper, gold, silver, or magnetic wires.
Blade: Predominantly angled
OAL: 115 mm
Catalog No. 50-193-1006 Supplier Electron Microscopy Sciences Supplier No. 7832510
Soft ESD safe ergonomic cushion grips (resistivity 10^8-10^9 Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Cutting tweezers for soft copper, gold, silver, or magnetic wires.
Blade: Predominantly angled
OAL: 115 mm