Learn More
Med Vet International Rubber Feeding Tubes, J0347E, 20Fr x 16in, 5 Count
Catalog No. 5000582027
Encompass_Preferred
20 Fr. 40cm (16") 5/pk. Small Animal Feeding Tube. Red latex rubber with two opposing lateral eyes. X-ray opaque. Sterile. Improved over red vinyl tubes because it is bio-compatible and allows more patient comfort with less rigidity. Can be reused and rea
Catalog No. 50-005-82027 Supplier Med Vet International Supplier No. JOR347E
20 Fr. 40cm (16") 5/pk. Small Animal Feeding Tube. Red latex rubber with two opposing lateral eyes. X-ray opaque. Sterile. Improved over red vinyl tubes because it is bio-compatible and allows more patient comfort with less rigidity. Can be reused and rea