Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laxco™ Rubber Grip for LMC-1000 Series Microscopes
SDP

Catalog No. LMCP0104792 Shop All Laxco Inc Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
LMCP0104792 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LMCP0104792 Supplier Laxco™ Supplier No. LMCP0104792

missing translation for 'validMainframeMsg'

Colored Hand Grips designed to be used with LMC-1000 Series Microscopes.

  • Choice any combination of colored handgrips for Laxco™ LMC-1000 series microscopes
  • Color code them based off from their use, classroom, school colors, or just your favorite color
  • Make your microscope unique, and more personalized
  • Hand grips are make of a soft, no-slip rubber, with finger indents to make carrying of your microscope easy and comfortable
  • Order them at the time of purchase of your microscope and we will install them before they ship, or add them to your existing microscope yourself
  • NOTE: for use with Laxco LMC-1000 biological series microscopes ONLY

Specifications

Color Green
For Use With (Equipment) LMC-1000 Series Microscopes
Material Rubber
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.