Learn More
Thomas Scientific Replacement silicone pipette holder
SDP
Catalog No. 501460561
Encompass
Designed for cordless work with glass or plastic pipettes Max. aspiration rate of 25 mL in 4.3 seconds Touch-control of aspiration rate Ergonomic handle Precision valve system The FastPette™ features high and low aspiration speeds and gravity or pump-supported dispensing settings. Safety valve prevents liquid from entering the device eliminating health hazards and damage to the device. Fully autoclavable nosepiece, pipette holder and filter help reduce cross-contamination. The NiMH rechargeable battery provides 8 hours of continuous use. Unit is fully operational during charging.
Catalog No. 50-146-0561 Supplier Thomas Scientific Supplier No. 1191X19
Designed for cordless work with glass or plastic pipettes Max. aspiration rate of 25 mL in 4.3 seconds Touch-control of aspiration rate Ergonomic handle Precision valve system The FastPette™ features high and low aspiration speeds and gravity or pump-supported dispensing settings. Safety valve prevents liquid from entering the device eliminating health hazards and damage to the device. Fully autoclavable nosepiece, pipette holder and filter help reduce cross-contamination. The NiMH rechargeable battery provides 8 hours of continuous use. Unit is fully operational during charging.