Learn More
Electron Microscopy Sciences Rubber Policeman | 12/Box
SDP
Catalog No. 501901993
Encompass_Preferred
Policeman is rubber cover to fit 5mm rods. The sealed end is cut at an angle to help stir at the bottom of container.
Catalog No. 50-190-1993 Supplier Electron Microscopy Sciences Supplier No. 6620712
Policeman is rubber cover to fit 5mm rods. The sealed end is cut at an angle to help stir at the bottom of container.