Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Rubber Shoe

Catalog No. 116002515 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24™ Instruments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-600-2515 FastPrep-24™ Instruments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-600-2515 Supplier MP Biomedicals™ Supplier No. 116002515
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Rubber Shoe is a FastPrep-24™ part/accessory.

  • Replacement part for FastPrep-24™ instruments.
TRUSTED_SUSTAINABILITY

Specifications

Material Rubber
For Use With (Equipment) FastPrep-24™ Instruments
Packaging Quantity 4 Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.