Learn More
MSC RUBBER SPLICE TAPE 23
Catalog No. 5000796943
Encompass
Highly conformable, self-fusing EPR (Ethylene Propylene Rubber) based tape for high-voltage splicing. Non-vulcanizing and shelf-stable with excellent electrical properties, suitable for insulation in low-voltage applications and splices up to 69,000 volts. Operates at emergency cable temperatures up to 266°F (130°C), maintaining properties whether stretched or unstretched.
  • Self-fusing Ethylene Propylene Rubber (EPR) based
  • Excellent electrical properties, high conformability
  • Physical and electrical properties unaffected by stretch
  • Primary electrical insulation for splicing cables up to 69,000 V
  • Operates up to 266°F (130°C) emergency temperature
  • Compatible with all solid dielectric cable insulation
  • Moisture sealing for electrical connections
  • Shelf life of 5 years
Catalog No. 50-007-96943 Supplier MSC Supplier No. 73664153
Highly conformable, self-fusing EPR (Ethylene Propylene Rubber) based tape for high-voltage splicing. Non-vulcanizing and shelf-stable with excellent electrical properties, suitable for insulation in low-voltage applications and splices up to 69,000 volts. Operates at emergency cable temperatures up to 266°F (130°C), maintaining properties whether stretched or unstretched.
  • Self-fusing Ethylene Propylene Rubber (EPR) based
  • Excellent electrical properties, high conformability
  • Physical and electrical properties unaffected by stretch
  • Primary electrical insulation for splicing cables up to 69,000 V
  • Operates up to 266°F (130°C) emergency temperature
  • Compatible with all solid dielectric cable insulation
  • Moisture sealing for electrical connections
  • Shelf life of 5 years