Learn More
Bellco Glass, Inc. Rubber STOPPER,#0
SDP
Supplier:  Bellco Glass, Inc. 192711110
Encompass_Preferred
A laboratory rubber stopper is mainly used in chemical laboratory in combination with flasks and test tub. Made from rubber Closure of flask Size: 0 Top dia: 17mm Bottom dia: 13.5mm Length: 25mm
Catalog No. 50-194-4700