Learn More
Chemglass Life Sciences Solid Rubber Stopper, #00, Black
SDP
Catalog No. 501531463
Encompass_Preferred
Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn. This item is RED rubber.
Catalog No. 50-153-1463 Supplier Chemglass Life Sciences Supplier No. CG302602
Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn. This item is RED rubber.