Learn More
Research Products International Corp Laboratory Grade Rubber Stopper, Black SBR Rubber, Size #000, 12.7 x 8.2 x 21mm, 183 per Package
SDP
Catalog No. 50998962
Encompass
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.
Catalog No. 50-998-962 Supplier Research Products International Corp Supplier No. 259150000
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.