Learn More
Chemglass Life Sciences Rubber Stopper, #000, One Hole, Black
SDP
Catalog No. 501531488
Encompass_Preferred
Rubber Stopper, #000, One Hole, Black. Similar to CG-3026 except with one hole. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (12.7mm), Bottom (8.2mm). Length: (21mm). Diameter of hole: (3mm).
Catalog No. 50-153-1488 Supplier Chemglass Life Sciences Supplier No. CG302701
Rubber Stopper, #000, One Hole, Black. Similar to CG-3026 except with one hole. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (12.7mm), Bottom (8.2mm). Length: (21mm). Diameter of hole: (3mm).