Learn More
Chemglass Life Sciences Solid Rubber Stopper, #1, Black
SDP
Catalog No. 501531465
Encompass_Preferred
Solid Rubber Stopper, #1, Black. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (19mm), Bottom (14mm). Length: (25mm).
Catalog No. 50-153-1465 Supplier Chemglass Life Sciences Supplier No. CG302604
Solid Rubber Stopper, #1, Black. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (19mm), Bottom (14mm). Length: (25mm).