Learn More
Chemglass Life Sciences Syringe, Polypropylene, Luer Lock, 20mL, Non-Sterile, Bulk
SDP
Catalog No. NC1188481
Encompass_Preferred
These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills.
Catalog No. NC1188481 Supplier Chemglass Life Sciences Supplier No. CG308304
These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills.