Learn More
Chemglass Life Sciences Rubber Stopper, #3, Two Holes, Black
SDP
Catalog No. 501531515
Encompass_Preferred
Rubber Stopper, #3, Two Holes, Black. Similar to CG-3026 and CG-3027 except with two holes. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (24mm), Bottom (18mm). Length: (25mm). Diameter of holes: (5mm).
Catalog No. 50-153-1515 Supplier Chemglass Life Sciences Supplier No. CG302805
Rubber Stopper, #3, Two Holes, Black. Similar to CG-3026 and CG-3027 except with two holes. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (24mm), Bottom (18mm). Length: (25mm). Diameter of holes: (5mm).