Learn More
Chemglass Life Sciences Rubber Stopper, #6-1/2, One Hole, Black
SDP
Catalog No. 501531497
Encompass_Preferred
Rubber Stopper, #6-1/2, One Hole, Black. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (34mm), Bottom (27mm). Length: (25mm). Diameter of hole: (5mm).
Catalog No. 50-153-1497 Supplier Chemglass Life Sciences Supplier No. CG302711
Rubber Stopper, #6-1/2, One Hole, Black. These stoppers are suitable for practically all general laboratory uses. Composition ensures resiliency and pliability over a long period and withstands laboratory conditions without hardening. The dimensions are in accordance with specifications established by the N.B.S. and are low in free sulfur (1/2%). Autoclavable at 250F for 30 minutes under water or steam conditions only (not dry heat). End user must test for their specific application. Diameter: Top (34mm), Bottom (27mm). Length: (25mm). Diameter of hole: (5mm).