Learn More
Research Products International Corp Laboratory Grade Rubber Stopper, Black SBR Rubber, Size #6, 32 x 26 x 25mm, 18 per Package
SDP
Catalog No. 50998974
Encompass
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA accordinig to standard industry size specification. Packaged in approxinmately one pound packages.
Catalog No. 50-998-974 Supplier Research Products International Corp Supplier No. 2591506
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA accordinig to standard industry size specification. Packaged in approxinmately one pound packages.