Learn More
Research Products International Corp Laboratory Grade Rubber Stopper, Black SBR Rubber, Size #8, 45 x 37 x 25mm, 12 per Package
SDP
Catalog No. 50998977
Encompass
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.
Catalog No. 50-998-977 Supplier Research Products International Corp Supplier No. 2591508
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.