Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Rubber Stoppers

Catalog No. 111020929 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Rubber Stopper
Material:
Butyl Rubber
Rubber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
11-102-0929 Rubber Stopper Butyl Rubber
11-100-4763 Rubber Stopper Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0929 Supplier MilliporeSigma™ Supelco™ Supplier No. 27232
Add to Cart
Add to Cart

Generally used to seal samples in vials

TRUSTED_SUSTAINABILITY

Specifications

Material Butyl Rubber
Color Gray
Product Type Rubber Stopper
For Use With (Application) Sealing Samples in Vials
Quantity 100/Pk.
Diameter (Metric) 20 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.