Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasticoid One-hole Rubber Stoppers for Kjeldahl Flasks
SDP

Catalog No. 14149B Shop All Plasticoid Products
Change view
Click to view available options
Color:
Light Tan
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-149B Light Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-149B Supplier Plasticoid Supplier No. U500M181
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Light tan rubber stoppers

  • One-hole style can be enlarged or the solid stopper bored to take larger sizes
  • Top diameter: 32mm; bottom diameter: 26mm; height: 34mm; one-hole bore diameter, 8mm

Specifications

Product Type Stopper (1 hole)
Diameter (Metric) Top 3.2 cm
Diameter (Metric) Bottom 2.6 cm
Total Positions 1
Height (Metric) 3.4 cm
Material Rubber
For Use With (Application) Food grade applications such as homebrew beer and wine making used with carboys
Color Light Tan
For Use With (Equipment) Kjeldahl flask
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.