Learn More
GSC Go Science Crazy Rubberized Cloth Apron
SCI_ED
Catalog No. p-7070796 Shop All GSC Products
Click to view available options
Dimensions (L x W):
24 x 30 in.
27 x 36 in.
27 x 42 in.
36 x 42 in.
36 x 46 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S13709 36 x 42 in.
S13710 36 x 46 in.
S13708 27 x 42 in.
S13707 27 x 36 in.
S13706 24 x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Intended for ages 13 and up. May contain latex.
Catalog No. S13709 Supplier GSC Go Science Crazy Supplier No. 1276310
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for ages 13 and up. May contain latex.
Provides basic protection from chemicals in laboratory or classroom settings. The GSC International Rubberized Cloth Apron is made of rubberized cloth and available in several sizes.
  • Great for laboratory and classroom safety
  • Material is rubberized and resistant to most common chemicals
  • Cloth tie strings provide steadiness and flexibility
  • Available in several sizes
Specifications
Material Rubberized Cloth
Includes Cloth tie strings
For Use With (Application) Laboratory or classroom settings
Dimensions (L x W) 36 x 42 in.
WARNING: Cancer - www.P65Warnings.ca.gov