Learn More
Sigma Aldrich Fine Chemicals Biosciences Rubberized square weighing
Catalog No. 502901334
Encompass
Rubberized polystyrene weighing boats with a smooth surface to provide accurate pour-outs with minimal sample loss. Shallow shape with a wide flat bottom to resist tipping. Rounded corners simplify transfer. Biologically inert and resistant to dilute and weak acids aqueous SOLUTIONons alcohols and bases. May be used with microwave moisture analyzers. Resist temperatures to 200Af (93AC).
Catalog No. 50-290-1334 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z708550500EA
Rubberized polystyrene weighing boats with a smooth surface to provide accurate pour-outs with minimal sample loss. Shallow shape with a wide flat bottom to resist tipping. Rounded corners simplify transfer. Biologically inert and resistant to dilute and weak acids aqueous SOLUTIONons alcohols and bases. May be used with microwave moisture analyzers. Resist temperatures to 200Af (93AC).