Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Brute Round Dolly, 350 Lb. Load Cap, Black
SDP
Catalog No. 5000790791
Encompass_Preferred
Easy twist-on, twist-off dolly provides safe mobility and handling of heavy loads. Five rugged casters help keep fully loaded Brute containers stable, even on rough and uneven floors, and swivel smoothly for easy maneuvering. Brute Structural Foam Dolly features a 350 lb. maximum load capacity and fits Rubbermaid 2620, 2632, 2643 and 2655 (sold separately).
Catalog No. 50-007-90791 Supplier Enterprise Technology Solutions Supplier No. RCP264000BK
Easy twist-on, twist-off dolly provides safe mobility and handling of heavy loads. Five rugged casters help keep fully loaded Brute containers stable, even on rough and uneven floors, and swivel smoothly for easy maneuvering. Brute Structural Foam Dolly features a 350 lb. maximum load capacity and fits Rubbermaid 2620, 2632, 2643 and 2655 (sold separately).