Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Brute Square Dolly, f/28/40/50 Gal, 17-1/4"x6-1/4", Black
SDP
Catalog No. 5000790754
Encompass_Preferred
Brute Square Dolly delivers easy mobility and maneuverability of heavy loads for Brute Square 28 gallon and 48 gallon Containers without lids (3526, 3536). All plastic, professional-grade construction resists rust, chipping and denting. Heavy-duty wheels with swivel capabilities are designed for ease of mobility. Dolly offers up to a 300 lb. continuous static load rating.
Catalog No. 50-007-90754 Supplier Enterprise Technology Solutions Supplier No. RCP353000BK
Brute Square Dolly delivers easy mobility and maneuverability of heavy loads for Brute Square 28 gallon and 48 gallon Containers without lids (3526, 3536). All plastic, professional-grade construction resists rust, chipping and denting. Heavy-duty wheels with swivel capabilities are designed for ease of mobility. Dolly offers up to a 300 lb. continuous static load rating.