Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Brute Step-On Rollout Cont, 65Gal, Gray
SDP
Catalog No. 5000790694
Encompass_Preferred
BRUTE Step-On Rollout Container is designed to make managing facility waste easier. With one step, the container's step-on mechanism lifts the lid and allows hands-free waste disposal. Heavy-duty wheels with high-performance treads are optimized for use indoors and outdoors. Reinforced rim delivers superior structural integrity, and robust lid is designed for added strength. Rollout lid opens fully and stays in place to prevent swinging during transport. 65-gallon container is compliant with ANSI Standard Z245.60 (Types B and G) for compatibility with auto-lifters. Smooth contours and seamless construction make cleaning easier and quicker to improve staff productivity. Ergonomic rear handle allows comfortable push or pull operation. High-quality materials plus a UV inhibitor extend the life of the container. BRUTE Step-On Rollout is designed to maintain performance at freezing temperatures as low as zero degrees Fahrenheit.
Catalog No. 50-007-90694 Supplier Enterprise Technology Solutions Supplier No. RCP1971968
BRUTE Step-On Rollout Container is designed to make managing facility waste easier. With one step, the container's step-on mechanism lifts the lid and allows hands-free waste disposal. Heavy-duty wheels with high-performance treads are optimized for use indoors and outdoors. Reinforced rim delivers superior structural integrity, and robust lid is designed for added strength. Rollout lid opens fully and stays in place to prevent swinging during transport. 65-gallon container is compliant with ANSI Standard Z245.60 (Types B and G) for compatibility with auto-lifters. Smooth contours and seamless construction make cleaning easier and quicker to improve staff productivity. Ergonomic rear handle allows comfortable push or pull operation. High-quality materials plus a UV inhibitor extend the life of the container. BRUTE Step-On Rollout is designed to maintain performance at freezing temperatures as low as zero degrees Fahrenheit.