Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Cotton Pad, f/Wall Washer Kit, Washable, WE
SDP
Catalog No. 5000790824
Encompass_Preferred
Replacement pads are designed for use in the Rubbermaid Commercial Wall Washer Kit. When attached to the plastic head on the wood or aluminum handle (all sold separately), the pads help you scrub walls clean. Raised-ribbed design works with either side-pressure or downward pressure mop wringers without requiring additional adapters. Washable cotton pads remove easily for convenient laundering.
Catalog No. 50-007-90824 Supplier Enterprise Technology Solutions Supplier No. RCPS299
Replacement pads are designed for use in the Rubbermaid Commercial Wall Washer Kit. When attached to the plastic head on the wood or aluminum handle (all sold separately), the pads help you scrub walls clean. Raised-ribbed design works with either side-pressure or downward pressure mop wringers without requiring additional adapters. Washable cotton pads remove easily for convenient laundering.