Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Duster, w/Frame, Microfiber, Quick-Connect, 22", WE
SDP
Catalog No. 5000790762
Encompass_Preferred
Part of the Rubbermaid Commercial Executive Series Hygen Cleaning System, Microfiber Quick-Connect Flexi-Wand and Duster connects with a telescoping handle (sold separately) to easily clean vents, furniture, bed covers, uniquely shaped fixtures, and more. It easily bends and conforms to irregular shapes to allow you to clean crown molding, tops of cabinets and other areas. It easily collects dust, hair and larger debris. Duster works great for cleaning furniture and bed covers. The Quick-Connect mechanism allows the wand to be used with any Rubbermaid Commercial telescoping handle. Washable microfiber sleeve is easy to replace by just sliding it off the wand. Use it wet or dry to trap the largest amount of dust and dirt. Replacement sleeves are sold separately for added product life.
Catalog No. 50-007-90762 Supplier Enterprise Technology Solutions Supplier No. RCPQ852WHI
Part of the Rubbermaid Commercial Executive Series Hygen Cleaning System, Microfiber Quick-Connect Flexi-Wand and Duster connects with a telescoping handle (sold separately) to easily clean vents, furniture, bed covers, uniquely shaped fixtures, and more. It easily bends and conforms to irregular shapes to allow you to clean crown molding, tops of cabinets and other areas. It easily collects dust, hair and larger debris. Duster works great for cleaning furniture and bed covers. The Quick-Connect mechanism allows the wand to be used with any Rubbermaid Commercial telescoping handle. Washable microfiber sleeve is easy to replace by just sliding it off the wand. Use it wet or dry to trap the largest amount of dust and dirt. Replacement sleeves are sold separately for added product life.