Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Finish Wet Mop, Antimicrobial, 24oz, 1" Headband, WE
SDP
Catalog No. 5000829516
Encompass_Preferred
Web Foot Finish Wet Mop is made from four-ply, looped-end synthetic yarn that is designed for a controlled release of floor finish. Tailband is sewn on the inside of mop and helps reduce the "drag" patterns common to most finish mops. It allows the mop to cover more floor area per stroke, saving time and money. Wet mop contains antimicrobial product protection that inhibits the growth of bacteria that cause odors and stains. Looped-end, open-twist yarn reduces linting. Mop is launderable up to 74 launderings.
Catalog No. 50-008-29516 Supplier Enterprise Technology Solutions Supplier No. RCPA413
Web Foot Finish Wet Mop is made from four-ply, looped-end synthetic yarn that is designed for a controlled release of floor finish. Tailband is sewn on the inside of mop and helps reduce the "drag" patterns common to most finish mops. It allows the mop to cover more floor area per stroke, saving time and money. Wet mop contains antimicrobial product protection that inhibits the growth of bacteria that cause odors and stains. Looped-end, open-twist yarn reduces linting. Mop is launderable up to 74 launderings.