Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Hamper, f/Linens, Mobile, Poly, 30 Gal Cap, Black
SDP
Catalog No. 5000790788
Encompass_Preferred
Premium Hamper is ideal for areas where medical waste originates, including patient rooms, lab procedure rooms and emergency rooms. It assists in complying with OSHA regulation; 29 CFR 1910.1030 "Occupational Exposure to Bloodborne Pathogens." Quiet, heavy-duty pedal allows hands-free sanitary use with a 30-Gallon Linen Hamper Bag (6350) and Can Liner (both sold separately). Slow-close integrated lid damper helps keep odors and bacteria from escaping. Metal bag straps keep can liners securely in place, eliminating the need to tie knots in can liners. Hamper includes two swivel and two fixed, lockable Quiet Casters for reduced operational noise in noise-sensitive environments. Construction of impact-resistant plastic and metal provides a long product life and easy maneuverability.
Catalog No. 50-007-90788 Supplier Enterprise Technology Solutions Supplier No. RCP6300BLA
Premium Hamper is ideal for areas where medical waste originates, including patient rooms, lab procedure rooms and emergency rooms. It assists in complying with OSHA regulation; 29 CFR 1910.1030 "Occupational Exposure to Bloodborne Pathogens." Quiet, heavy-duty pedal allows hands-free sanitary use with a 30-Gallon Linen Hamper Bag (6350) and Can Liner (both sold separately). Slow-close integrated lid damper helps keep odors and bacteria from escaping. Metal bag straps keep can liners securely in place, eliminating the need to tie knots in can liners. Hamper includes two swivel and two fixed, lockable Quiet Casters for reduced operational noise in noise-sensitive environments. Construction of impact-resistant plastic and metal provides a long product life and easy maneuverability.