Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Handle, f/Wet Mops, Clamp Style, Aluminum, 60", YW/SR
SDP
Catalog No. 5000790848
Encompass_Preferred
Gripper Wet Mop Handle makes removing and replacing mopheads easy and efficient. Jaws of the mop handle hold mopheads firmly. This clamp-style handle should be used on wet mopheads with 5" (12.7 cm) headbands only (sold separately). The head of the frame is made of plastic. Handle is made of aluminum.
Catalog No. 50-007-90848 Supplier Enterprise Technology Solutions Supplier No. RCPH226
Gripper Wet Mop Handle makes removing and replacing mopheads easy and efficient. Jaws of the mop handle hold mopheads firmly. This clamp-style handle should be used on wet mopheads with 5" (12.7 cm) headbands only (sold separately). The head of the frame is made of plastic. Handle is made of aluminum.