Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products HYGEN™ R-40 Recycled Microfiber Cloths, 16"x16", 24/BG, Red
SDP
Catalog No. 5000829886
Encompass_Preferred
HYGEN™ R-40 Recycled Microfiber Cleaning Cloths deliver sustainable strength and proven performance for every cleaning task. Made with 40% post-consumer recycled polyester, HYGEN™ cloth repurposes plastic bottles to reduce reliance on virgin plastics. Engineered for superior durability, it withstands 200 commercial launderings (100 with bleach) while maintaining bright color for easy color-coding. Color stays bright and bold for the entire life of the product. Advanced warp-knit microfiber is split into 16 filaments with over 95% splitting ratio, creating maximum surface area to trap dirt and remove 99.9% of tested fungi and bacteria using only water. Designed for wet or dry use, these color-coded cloths help prevent cross-contamination and drive higher productivity across various cleaning tasks. From commercial facilities to home cleaning, HYGEN™ R-40 is built to perform, proving that cleaning supplies can be powerful and responsible.
Catalog No. 50-008-29886 Supplier Enterprise Technology Solutions Supplier No. RCP1820581
HYGEN™ R-40 Recycled Microfiber Cleaning Cloths deliver sustainable strength and proven performance for every cleaning task. Made with 40% post-consumer recycled polyester, HYGEN™ cloth repurposes plastic bottles to reduce reliance on virgin plastics. Engineered for superior durability, it withstands 200 commercial launderings (100 with bleach) while maintaining bright color for easy color-coding. Color stays bright and bold for the entire life of the product. Advanced warp-knit microfiber is split into 16 filaments with over 95% splitting ratio, creating maximum surface area to trap dirt and remove 99.9% of tested fungi and bacteria using only water. Designed for wet or dry use, these color-coded cloths help prevent cross-contamination and drive higher productivity across various cleaning tasks. From commercial facilities to home cleaning, HYGEN™ R-40 is built to perform, proving that cleaning supplies can be powerful and responsible.