Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Mobile Utility Cart,300 lb. Cap,15-1/8"x20-3/4"x36",Black
SDP
Catalog No. 5000791937
Encompass_Preferred
Three-Shelf Utility Cart is ideal for moving equipment from office to conference room. Design features tough, structural web construction; aluminum frame with 4" nonmarking casters and a large 20" x 40" three-shelf work area. Textured surface resists scratching. Utility cart holds up to 300 lb.
Catalog No. 50-007-91937 Supplier Enterprise Technology Solutions Supplier No. RCP409100BK
Three-Shelf Utility Cart is ideal for moving equipment from office to conference room. Design features tough, structural web construction; aluminum frame with 4" nonmarking casters and a large 20" x 40" three-shelf work area. Textured surface resists scratching. Utility cart holds up to 300 lb.