Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Pad,f/Adaptable Flat Mop,Microfiber,18"W,Blue
SDP
Catalog No. 5000790852
Encompass_Preferred
Adaptable Flat Mop Pad helps improve cleaning efficacy and connects to the Adaptable Flat Mop Frame (not included) via tabs or pockets for two styles of microfiber mopping in large spaces. Microfiber pad removes 99.7 percent or more of tested viruses and bacteria with water only to help improve cleaning efficacy. These tested germs include Feline Calicivirus (surrogate for Norovirus), MRSA, Pseudo and C. diff. Microfiber mop pad is 70 percent lighter when fully soaked vs. a traditional string mop. Convenient clips help avoid contact with soiled mop pad during disposal. You can mop floors 15 percent faster and floors dry 4 times faster when mopping with Adaptable Flat Mop microfiber pads, compared to RCP large cotton string mops. Pad has been tested to withstand up to 200 commercial launderings and 100 launderings with bleach.
Catalog No. 50-007-90852 Supplier Enterprise Technology Solutions Supplier No. RCP2132427
Adaptable Flat Mop Pad helps improve cleaning efficacy and connects to the Adaptable Flat Mop Frame (not included) via tabs or pockets for two styles of microfiber mopping in large spaces. Microfiber pad removes 99.7 percent or more of tested viruses and bacteria with water only to help improve cleaning efficacy. These tested germs include Feline Calicivirus (surrogate for Norovirus), MRSA, Pseudo and C. diff. Microfiber mop pad is 70 percent lighter when fully soaked vs. a traditional string mop. Convenient clips help avoid contact with soiled mop pad during disposal. You can mop floors 15 percent faster and floors dry 4 times faster when mopping with Adaptable Flat Mop microfiber pads, compared to RCP large cotton string mops. Pad has been tested to withstand up to 200 commercial launderings and 100 launderings with bleach.