Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Thermometer, f/Freezer/Refrigerators, -20 to 80F, Silver
SDP
Catalog No. 5000828048
Encompass_Preferred
Thermometer helps monitor the temperatures in your refrigerator or freezer. It is easy to install. Simply hang or set on a shelf. Clearly marked dial is easy to read. It includes both Fahrenheit and Celsius graduations. Thermometer ranges from negative 20 to 80 degrees Fahrenheit (negative 28.9 to 26.7 degrees Celsius).
Catalog No. 50-008-28048 Supplier Enterprise Technology Solutions Supplier No. RCPPELR80DC
Thermometer helps monitor the temperatures in your refrigerator or freezer. It is easy to install. Simply hang or set on a shelf. Clearly marked dial is easy to read. It includes both Fahrenheit and Celsius graduations. Thermometer ranges from negative 20 to 80 degrees Fahrenheit (negative 28.9 to 26.7 degrees Celsius).