Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Tilt Truck,Utility Duty,450 lb Cap,28"x56-4/5"x38-3/5",BK
SDP
Catalog No. 5000829888
Encompass_Preferred
Tilt Dump Truck offers industrial-strength construction to easily transport heavy loads up to 450 lb. 5" swivel casters and 12" inset wheels provide smooth handling and maneuverability. It is easy to maneuver through commercial doorways and onto elevators. A constant force design allows controlled dumping by one person. Smooth surface is easy-to-clean and resists pitting and corroding. The 1/2 cubic yard dump truck features a powder-coated steel frame for corrosion-resistance, durable rotomolded plastic and leakproof, polyethylene bottom body.
Catalog No. 50-008-29888 Supplier Enterprise Technology Solutions Supplier No. RCP130400BK
Tilt Dump Truck offers industrial-strength construction to easily transport heavy loads up to 450 lb. 5" swivel casters and 12" inset wheels provide smooth handling and maneuverability. It is easy to maneuver through commercial doorways and onto elevators. A constant force design allows controlled dumping by one person. Smooth surface is easy-to-clean and resists pitting and corroding. The 1/2 cubic yard dump truck features a powder-coated steel frame for corrosion-resistance, durable rotomolded plastic and leakproof, polyethylene bottom body.