Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Utility Cart, 200 lb. Cap, 33-5/8"x18-5/8"x37-3/4", PM
SDP
Catalog No. 5000791977
Encompass_Preferred
Three-shelf, utility cart is versatile for audiovisual, video, filing and foodservice purposes and offers a high-style appearance with brushed aluminum uprights. Smooth surfaces are easy to clean. Quiet casters and plastic construction help eliminate noise. Shelves feature a 1/4" lip on three sides and one flat side for easy loading. Cart holds a total of up to 200 lb. Maneuver cart easily with dual-molded handles and 4" nonmarking swivel casters.
Catalog No. 50-007-91977 Supplier Enterprise Technology Solutions Supplier No. RCP342488PM
Three-shelf, utility cart is versatile for audiovisual, video, filing and foodservice purposes and offers a high-style appearance with brushed aluminum uprights. Smooth surfaces are easy to clean. Quiet casters and plastic construction help eliminate noise. Shelves feature a 1/4" lip on three sides and one flat side for easy loading. Cart holds a total of up to 200 lb. Maneuver cart easily with dual-molded handles and 4" nonmarking swivel casters.