Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Waste Container,Step-on,23 Gal,19-7/8"x16-1/2"x32-3/5",Beige
SDP
Catalog No. 5000829559
Encompass_Preferred
Mobile Step-on Container features a tight-fitting, overlapping lid to contain odors and is approved for fire safety. Slow-close, integrated lid damper helps keep odors and bacteria from escaping. All-plastic construction is tough and puncture-resistant. Heavy-duty pedal controls entry and delivers hands-free sanitary use. Factory Mutual (FM) approved step-on container assists in complying with OSHA regulation; 29 CFR 1910.1030 Occupational Exposure to Bloodborne Pathogens. Capacity is 23 gallons.
Catalog No. 50-008-29559 Supplier Enterprise Technology Solutions Supplier No. RCP614600BG
Mobile Step-on Container features a tight-fitting, overlapping lid to contain odors and is approved for fire safety. Slow-close, integrated lid damper helps keep odors and bacteria from escaping. All-plastic construction is tough and puncture-resistant. Heavy-duty pedal controls entry and delivers hands-free sanitary use. Factory Mutual (FM) approved step-on container assists in complying with OSHA regulation; 29 CFR 1910.1030 Occupational Exposure to Bloodborne Pathogens. Capacity is 23 gallons.