Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Wastebasket,Fire-resistant,14 Qt,8-1/4"x11-1/8"x12-1/4",BK
SDP
Catalog No. 5000828894
Encompass_Preferred
Fire-resistant wastebasket can contain burning paper without the wastebasket melting or contributing fuel to the container's contents. Textured finish helps minimize appearance of surface scratches. Long-lasting fiberglass resists chips and dents. It won't rust. This 14-quart wastebasket offers a contemporary shape and UL rating and is ideal for industrial or hospital use.
Catalog No. 50-008-28894 Supplier Enterprise Technology Solutions Supplier No. RCP254100BK
Fire-resistant wastebasket can contain burning paper without the wastebasket melting or contributing fuel to the container's contents. Textured finish helps minimize appearance of surface scratches. Long-lasting fiberglass resists chips and dents. It won't rust. This 14-quart wastebasket offers a contemporary shape and UL rating and is ideal for industrial or hospital use.