Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Wastebasket,Fire-Resistant,28 Qt,15.25"x14.5"x10.5",BG
SDP
Catalog No. 5000828876
Encompass_Preferred
Fire-resistant wastebasket can contain burning paper without the wastebasket melting or contributing fuel to the container's contents. Textured finish helps minimize appearance of surface scratches. Thermoset polyester resists heat and impacts. It won't rust. This 28-quart wastebasket offers a contemporary shape and UL rating and is ideal for industrial or hospital use.
Catalog No. 50-008-28876 Supplier Enterprise Technology Solutions Supplier No. RCP254300BG
Fire-resistant wastebasket can contain burning paper without the wastebasket melting or contributing fuel to the container's contents. Textured finish helps minimize appearance of surface scratches. Thermoset polyester resists heat and impacts. It won't rust. This 28-quart wastebasket offers a contemporary shape and UL rating and is ideal for industrial or hospital use.