Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Wet Pad, Microfiber, Hygen, ZigZag, 18", Green
SDP
Catalog No. 5000829585
Encompass_Preferred
HYGEN™ 18" Microfiber Wet Pad removes 99.9% or more of tested fungi, viruses and bacteria with water alone. Combining superior microfiber with a built-in scrubber, HYGEN™ Microfiber helps prevent cross-transmission and reduces the risk of healthcare-associated infections. It helps reduce cross-contamination with color-coded edge tape that encourages cleaning with different colors by area or task. Patented zig-zag design is made of polyester and polypropylene for added scrubbing power. Optimized face material construction balances drag force and cleaning efficacy for an effortless experience. Absorbent inner layers help hold enough solution to clean up to 250 square feet. Industry-leading loop backing is made of 100% polyester and securely attaches to the HYGEN™ 17" Quick Connect Frame (not included). Wet pad is tested to withstand 200 launderings to CDC guidelines in hot water (160 degrees Fahrenheit) with chlorine bleach.
Catalog No. 50-008-29585 Supplier Enterprise Technology Solutions Supplier No. RCPQ410GRN
HYGEN™ 18" Microfiber Wet Pad removes 99.9% or more of tested fungi, viruses and bacteria with water alone. Combining superior microfiber with a built-in scrubber, HYGEN™ Microfiber helps prevent cross-transmission and reduces the risk of healthcare-associated infections. It helps reduce cross-contamination with color-coded edge tape that encourages cleaning with different colors by area or task. Patented zig-zag design is made of polyester and polypropylene for added scrubbing power. Optimized face material construction balances drag force and cleaning efficacy for an effortless experience. Absorbent inner layers help hold enough solution to clean up to 250 square feet. Industry-leading loop backing is made of 100% polyester and securely attaches to the HYGEN™ 17" Quick Connect Frame (not included). Wet pad is tested to withstand 200 launderings to CDC guidelines in hot water (160 degrees Fahrenheit) with chlorine bleach.