Learn More
MSC Rubbermaid Cube Truck, Polyethylene, 12 cu. ft., 400 lb. Capacity, 1 ea
Catalog No. 5000514969
Encompass
This bulk cube truck is a versatile material handling solution designed for the efficient transport of waste, laundry, linens, and supplies. Constructed from durable, rotationally molded polyethylene with a supportive metal frame, it provides long-lasting performance in demanding environments. The truck features a 12 cu. ft. volume capacity and is rated for a load capacity of up to 400 lb.
  • Diamond wheel pattern for improved maneuverability
  • Smooth surfaces for easy cleaning and maintenance
  • Integrated rib structure on the rim to increase strength and resist crushing
  • Reinforced base engineered to withstand dragging over rough surfaces
  • Compatible with optional domed lids to conceal loads without reducing capacity
  • Commercial-grade construction with UV inhibitor for extended service life
  • Compliant with USDA Meat and Poultry Equipment Group guidelines
Catalog No. 50-005-14969 Supplier MSC Supplier No. 81746810
This bulk cube truck is a versatile material handling solution designed for the efficient transport of waste, laundry, linens, and supplies. Constructed from durable, rotationally molded polyethylene with a supportive metal frame, it provides long-lasting performance in demanding environments. The truck features a 12 cu. ft. volume capacity and is rated for a load capacity of up to 400 lb.
  • Diamond wheel pattern for improved maneuverability
  • Smooth surfaces for easy cleaning and maintenance
  • Integrated rib structure on the rim to increase strength and resist crushing
  • Reinforced base engineered to withstand dragging over rough surfaces
  • Compatible with optional domed lids to conceal loads without reducing capacity
  • Commercial-grade construction with UV inhibitor for extended service life
  • Compliant with USDA Meat and Poultry Equipment Group guidelines